Terephthalonitrile ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 1,4-Benzenedicarbonitrile, Terephthalodinitrile, Benzene-1,4-dicarbonitrile, 1,4-Dicyanobenzene

B23137.14 B23137.22
ALFAB23137.14EA 69.6 SGD
ALFAB23137.14 ALFAB23137.22
Terephthalonitrile ≥98%
Terephthalonitrile

Purity: 98% CAS No.: 623-26-7 Molecular Formula: C8H4N2 Formula Weight: 128.13 Odour: Odourless Melting Point: 224-227° Beilstein Reference: 1072210 MDL No.: MFCD00001810

Formula: C₈H₄N₂
MW: 128,13 g/mol
Melting Pt: 224…227 °C
Density: 1,29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001810
CAS Number: 623-26-7
EINECS: 210-783-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR