Veratrohydrazide ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 3,4-Dimethoxybenzohydrazide, 3,4-Dimethoxybenzene-1-carbohydrazide, 3,4-Dimethoxybenzoic hydrazide, 3,4-Dimethoxybenzoic acid hydrazide, 3,4-Dimethoxybenzhydrazide, 3,4-Dimethoxybenzenecarbohydrazide

A10404.06 A10404.14
ALFAA10404.06EA 0 SGD
ALFAA10404.06 ALFAA10404.14
Veratrohydrazide ≥98%
Veratrohydrazide

Purity: 98+% CAS No.: 41764-74-3 Molecular Formula: C9H12N2O3 Formula Weight: 196.21 Melting Point: 143-146° Beilstein Reference: 2651843

Formula: C₉H₁₂N₂O₃
MW: 196,21 g/mol
Melting Pt: 143…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00017061
CAS Number: 41764-74-3
EINECS: 255-541-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR