2,3-Difluorophenylboronic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Dihydroxy(2,3-difluorophenyl)borane, 2,3-Difluorobenzeneboronic acid, (2,3-Difluorophenyl)metaboric acid
L18012.06
L18012.03
L18012.14
ALFAL18012.06EA
0
SGD
ALFAL18012.06
ALFAL18012.03
ALFAL18012.14
2,3-Difluorophenylboronic acid ≥98%
2,3-Difluorophenylboronic acid
Purity: 98%
CAS No.: 121219-16-7
Molecular Formula: C6H5BF2O2
Formula Weight: 157.91
Melting Point: ca 225° dec.
Beilstein Reference: 6594341
Formula:
C₆H₅BF₂O₂ MW: 157,91 g/mol Melting Pt: ∼ 225 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD01863170 CAS Number: 121219-16-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...