2-(Trifluoromethoxy)phenylboronic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Trifluoromethoxyphenylboronic acid, 2-(Trifluoromethoxy)benzeneboronic acid, (2-(Trifluoromethoxy)phenyl)metaboric acid, Dihydroxy(2-(trifluoromethoxy)phenyl)borane
L19774.MD
L19774.03
ALFAL19774.MDEA
0
SGD
ALFAL19774.MD
ALFAL19774.03
2-(Trifluoromethoxy)phenylboronic acid ≥98%
2-(Trifluoromethoxy)phenylboronic acid
Purity: 98%
CAS No.: 175676-65-0
Molecular Formula: C7H6BF3O3
Formula Weight: 205.93
Melting Point: 118-120°
Beilstein Reference: 8766167
Formula:
C₇H₆BF₃O₃ MW: 205,93 g/mol Melting Pt: 118…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD01320763 CAS Number: 175676-65-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...