3-Fluoro-4-methoxybenzoic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
B21921.03
B21921.06
ALFAB21921.03EA
0
SGD
ALFAB21921.03
ALFAB21921.06
3-Fluoro-4-methoxybenzoic acid ≥98%
3-Fluoro-4-methoxybenzoic acid
Purity: 98%
CAS No.: 403-20-3
Molecular Formula: C8H7FO3
Formula Weight: 170.14
Melting Point: 209-213°
Beilstein Reference: 2087583
Formula:
C₈H₇FO₃ MW: 170,14 g/mol Melting Pt: 209…213 °C Storage Temperature: Ambient |
MDL Number:
MFCD00060675 CAS Number: 403-20-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...