4-Fluoro-3-methoxybenzoic acid 95%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Methoxy-4-fluorobenzoic acid

L19673.03 L19673.06
ALFAL19673.03EA 0 SGD
ALFAL19673.03 ALFAL19673.06
4-Fluoro-3-methoxybenzoic acid 95%
4-Fluoro-3-methoxybenzoic acid

Purity: 95% CAS No.: 82846-18-2 Molecular Formula: C8H7FO3 Formula Weight: 170.14 Melting Point: 206-208° Beilstein Reference: 4178071

Formula: C₈H₇FO₃
MW: 170,14 g/mol
Melting Pt: 206…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00272134
CAS Number: 82846-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR