3,4-Difluorosalicylic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
H26088.MD
H26088.03
ALFAH26088.MDEA
0
SGD
ALFAH26088.MD
ALFAH26088.03
3,4-Difluorosalicylic acid ≥98%
3,4-Difluorosalicylic acid
Purity: 98%
CAS No.: 189283-51-0
Molecular Formula: C7H4F2O3
Formula Weight: 174.10
Melting Point: 177-179°
MDL No.: MFCD03428564
Formula:
C₇H₄F₂O₃ MW: 174,1 g/mol Melting Pt: 177…179 °C Storage Temperature: Ambient |
MDL Number:
MFCD03428564 CAS Number: 189283-51-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...