1-Iodo-3-(trifluoromethoxy)benzene ≥97%
Supplier: Thermo Scientific
|
Warning
|
B24169.03
B24169.06
ALFAB24169.03EA
0
SGD
ALFAB24169.03
ALFAB24169.06
1-Iodo-3-(trifluoromethoxy)benzene ≥97%
1-Iodo-3-(trifluoromethoxy)benzene
Purity: 97+%
CAS No.: 198206-33-6
Molecular Formula: C7H4F3IO
Formula Weight: 288.01
Flash Point: 65°(150°F)
Refractive Index: 1.5200
Boiling Point: 185-186°
MDL No.: MFCD01090992
Formula:
C₇H₄F₃IO Boiling Pt: 185…186 °C Density: 1,855 Flash Pt: 65 °C (150 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01090992 CAS Number: 198206-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...