2,4,6-Trifluorophenyl isocyanate tech. 85%, Technical Grade
Supplier: Thermo Scientific
|
Danger
|
L19349.03
L19349.06
ALFAL19349.03EA
195
SGD
ALFAL19349.03
ALFAL19349.06
2,4,6-Trifluorophenyl isocyanate tech. 85%, Technical Grade
2,4,6-Trifluorophenyl isocyanate
Purity: 85%
CAS No.: 50528-80-8
Molecular Formula: C7H2F3NO
Formula Weight: 173.11
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4760
Beilstein Reference: 6325663
UN No.: 3080
MDL No.: MFCD03426028
Formula:
C₇H₂F₃NO MW: 173,09 g/mol Boiling Pt: 184 °C (1013 hPa) Storage Temperature: Refrigerator |
MDL Number:
MFCD03426028 CAS Number: 50528-80-8 UN: 3080 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...