2,6-Difluorophenyl isocyanate ≥97%
Supplier: Thermo Scientific
|
Danger
|
L14779.03
L14779.06
ALFAL14779.03EA
131
SGD
ALFAL14779.03
ALFAL14779.06
2,6-Difluorophenyl isocyanate ≥97%
2,6-Difluorophenyl isocyanate
Purity: 97%
CAS No.: 65295-69-4
Molecular Formula: C7H3F2NO
Formula Weight: 155.11
Flash Point: 58°(136°F)
Refractive Index: 1.4928
Melting Point: 22-25°
Boiling Point: 60°/10mm
Beilstein Reference: 7115873
UN No.: 2930
MDL No.: MFCD00001998
Formula:
C₇H₃F₂NO MW: 155,1 g/mol Boiling Pt: 60 °C (10 mmHg) Melting Pt: 22…25 °C Flash Pt: 58 °C (136 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001998 CAS Number: 65295-69-4 UN: 2930 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...