4-Amino-2-fluorophenol 97%
Supplier: Thermo Scientific
|
Warning
|
H25936.06
H25936.03
ALFAH25936.06EA
774
SGD
ALFAH25936.06
ALFAH25936.03
4-Amino-2-fluorophenol 97%
4-Amino-2-fluorophenol
Purity: 97%
CAS No.: 399-96-2
Molecular Formula: C6H6FNO
Formula Weight: 127.12
Melting Point: 170-172°
MDL No.: MFCD00671760
Formula:
C₆H₆FNO MW: 127,12 g/mol Melting Pt: 170…172 °C Storage Temperature: Ambient |
MDL Number:
MFCD00671760 CAS Number: 399-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...