3,4,5-Trifluorobenzenesulphonyl chloride ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
3,4,5-Trifluorobenzene-1-sulphonyl chloride
L19905.03
L19905.06
ALFAL19905.03EA
0
SGD
ALFAL19905.03
ALFAL19905.06
3,4,5-Trifluorobenzenesulphonyl chloride ≥98%
3,4,5-Trifluorobenzenesulphonyl chloride
Purity: 98%
CAS No.: 351003-43-5
Molecular Formula: C6H2ClF3O2S
Formula Weight: 230.59
Flash Point:
Formula:
C₆H₂ClF₃O₂S MW: 230,59 g/mol Boiling Pt: ∼230 °C Density: 1,643 Flash Pt: >100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03094559 CAS Number: 351003-43-5 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...