4-Iodo-2-nitroanisole ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-Iodo-1-methoxy-2-nitrobenzene
B25666.03
B25666.06
ALFAB25666.03EA
83.2
SGD
ALFAB25666.03
ALFAB25666.06
4-Iodo-2-nitroanisole ≥98%
4-Iodo-2-nitroanisole
Purity: 98%
CAS No.: 52692-09-8
Molecular Formula: C7H6INO3
Formula Weight: 279.04
Melting Point: 96-98°
MDL No.: MFCD08060933
Formula:
C₇H₆INO₃ MW: 279,03 g/mol Melting Pt: 96…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD08060933 CAS Number: 52692-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...