2,5-Dimethoxyphenyl isocyanate ≥99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Isocyanato-1,4-dimethoxybenzene
L11381.06
L11381.03
L11381.14
ALFAL11381.06EA
180
SGD
ALFAL11381.06
ALFAL11381.03
ALFAL11381.14
2,5-Dimethoxyphenyl isocyanate ≥99%
2,5-Dimethoxyphenyl isocyanate
Purity: 99%
CAS No.: 56309-62-7
Molecular Formula: C9H9NO3
Formula Weight: 179.18
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₉H₉NO₃ MW: 179,18 g/mol Boiling Pt: 152…154 °C (26 mmHg) Melting Pt: 38…42 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002006 CAS Number: 56309-62-7 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...