3-Nitrophenyl isothiocyanate ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
1-Isothiocyanato-3-nitrobenzene, m-Nitrophenylisothiocyanat
L11335.03
ALFAL11335.03EA
66.5
SGD
ALFAL11335.03
3-Nitrophenyl isothiocyanate ≥97%
3-Nitrophenyl isothiocyanate
Purity: 97%
CAS No.: 3529-82-6
Molecular Formula: C7H4N2O2S
Formula Weight: 180.19
Odour: Irritating
Melting Point: 56-58°
Beilstein Reference: 2094061
UN No.: 2923
MDL No.: MFCD00041245
Formula:
C₇H₄N₂O₂S MW: 180.19 g/mol Boiling Pt: 275…280 °C (1013 hPa) Melting Pt: 56…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041245 CAS Number: 3529-82-6 UN: 2923 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...