3,5-Dimethoxyphenyl isothiocyanate ≥97%
Supplier: Thermo Scientific
Synonyms:
1,3-Dimethoxy-5-isothiocyanatobenzene
A15728.03
A15728.06
ALFAA15728.03EA
0
SGD
ALFAA15728.03
ALFAA15728.06
3,5-Dimethoxyphenyl isothiocyanate ≥97%
3,5-Dimethoxyphenyl isothiocyanate
Purity: 97%
CAS No.: 104968-58-3
Molecular Formula: C9H9NO2S
Formula Weight: 195.24
Melting Point: 49-52°
Boiling Point: 130-132°/3mm
UN No.: 2923
MDL No.: MFCD00041077
Formula:
C₉H₉NO₂S Boiling Pt: 130…132 °C (3 mmHg) Melting Pt: 51 °C |
MDL Number:
MFCD00041077 CAS Number: 104968-58-3 UN: 2923 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...