1,3-Dimethoxy-4-nitrobenzene ≥97%

Supplier: Thermo Scientific

Synonyms: 2,4-Dimethoxynitrobenzene, 2,4-Dimethoxy-1-nitrobenzene

B25353.06 B25353.14
ALFAB25353.06EA 0 SGD
ALFAB25353.06 ALFAB25353.14
1,3-Dimethoxy-4-nitrobenzene ≥97%
1,3-Dimethoxy-4-nitrobenzene

Purity: 97% CAS No.: 4920-84-7 Molecular Formula: C8H9NO4 Formula Weight: 183.16 Melting Point: 75-77° MDL No.: MFCD00024210

Formula: C₈H₉NO₄
MW: 183,16 g/mol
Melting Pt: 75…77 °C
Density: 1,1876 g/cm³ (132 °C)
Storage Temperature: Ambient
MDL Number: MFCD00024210
CAS Number: 4920-84-7
EINECS: 225-551-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR