1-Bromo-4-nonylbenzene ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (4-Bromophenyl)nonane

H27042.03 H27042.09
ALFAH27042.03EA 76.6 SGD
ALFAH27042.03 ALFAH27042.09
1-Bromo-4-nonylbenzene ≥98%
1-Bromo-4-nonylbenzene

Purity: 98% CAS No.: 51554-94-0 Molecular Formula: C15H23Br Formula Weight: 283.25 Flash Point: 120°(248°F) Refractive Index: 1.509 Boiling Point: 107°/0.2mm MDL No.: MFCD00061116

Formula: C₁₅H₂₃Br
Boiling Pt: 107 °C (0,2 mmHg)
Density: 1,115
Flash Pt: >120 °C (248 °F)
Storage Temperature: Ambient
MDL Number: MFCD00061116
CAS Number: 51554-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR