α-Bromo-3,4-dichlorotoluene ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
3,4-Dichlorobenzyl bromide
L05254.06
L05254.14
ALFAL05254.06EA
106
SGD
ALFAL05254.06
ALFAL05254.14
α-Bromo-3,4-dichlorotoluene ≥98%
α-Bromo-3,4-dichlorotoluene
Purity: 98%
CAS No.: 18880-04-1
Molecular Formula: C7H5BrCl2
Formula Weight: 239.93
Flash Point:
Formula:
C₇H₅BrCl₂ MW: 239,93 g/mol Boiling Pt: 116…119 °C (1 mmHg) Density: 1,65 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041363 CAS Number: 18880-04-1 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...