4-Bromo-2-chlorotoluene ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-Bromo-3-chloro-4-methylbenzene
B22293.14
B22293.22
ALFAB22293.14EA
0
SGD
ALFAB22293.14
ALFAB22293.22
4-Bromo-2-chlorotoluene ≥98%
4-Bromo-2-chlorotoluene
Purity: 98%
CAS No.: 89794-02-5
Molecular Formula: C7H6BrCl
Formula Weight: 205.48
Flash Point: 104°(219°F)
Refractive Index: 1.5735
Boiling Point: 212°
Beilstein Reference: 3234853
MDL No.: MFCD00060651
Formula:
C₇H₆BrCl MW: 205,48 g/mol Boiling Pt: 212 °C Density: 1,538 Flash Pt: 104 °C (219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00060651 CAS Number: 89794-02-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...