4'-Bromopropiophenone ≥97%

Supplier: Thermo Scientific
Warning

A14349.09 A14349.14 A14349.18 A14349.30
ALFAA14349.09EA 0 SGD
ALFAA14349.09 ALFAA14349.14 ALFAA14349.18 ALFAA14349.30
4'-Bromopropiophenone ≥97%
4'-Bromopropiophenone

Purity: 97% CAS No.: 10342-83-3 Molecular Formula: C9H9BrO Formula Weight: 213.08 Flash Point: 110°(230°F) Melting Point: 45-47° Boiling Point: 129-131°/7mm Beilstein Reference: 1210311 MDL No.: MFCD00000106

Formula: C₉H₉BrO
Boiling Pt: 129…131 °C (7 mmHg)
Melting Pt: 45…47 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000106
CAS Number: 10342-83-3
EINECS: 233-745-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR