2-Bromo-6-fluorotoluene ≥98%
Supplier: Thermo Scientific
|
Warning
|
L07511.09
L07511.04
ALFAL07511.09EA
606
SGD
ALFAL07511.09
ALFAL07511.04
2-Bromo-6-fluorotoluene ≥98%
2-Bromo-6-fluorotoluene
Purity: 98%
CAS No.: 1422-54-4
Molecular Formula: C7H6BrF
Formula Weight: 189.03
Flash Point: 76°(168°F)
Refractive Index: 1.5330
Boiling Point: 180°
Beilstein Reference: 2433658
MDL No.: MFCD00051438
Formula:
C₇H₆BrF MW: 189,03 g/mol Boiling Pt: 180 °C Density: 1,53 Flash Pt: 76 °C (168 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051438 CAS Number: 1422-54-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...