2-(Bromomethyl)-1-fluoro-3-nitrobenzene ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 2-Fluoro-6-nitrobenzyl bromide, 1-(Bromomethyl)-2-fluoro-6-nitrobenzene

L16240.03 L16240.06
ALFAL16240.03EA 87.9 SGD
ALFAL16240.03 ALFAL16240.06
2-(Bromomethyl)-1-fluoro-3-nitrobenzene ≥98%
2-(Bromomethyl)-1-fluoro-3-nitrobenzene

Purity: 98+% CAS No.: 1958-93-6 Molecular Formula: C7H5BrFNO2 Formula Weight: 234.02 Odour: Pungent Melting Point: 50-54° Beilstein Reference: 2503670 UN No.: 3261 MDL No.: MFCD00042918

Formula: C₇H₅BrFNO₂
MW: 234,02 g/mol
Melting Pt: 50…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00042918
CAS Number: 1958-93-6
EINECS: 217-801-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR