2-Bromo-5-iodotoluene ≥98%
Supplier: Thermo Scientific
|
Warning
|
B22462.09
B22462.88
ALFAB22462.09EA
0
SGD
ALFAB22462.09
ALFAB22462.88
2-Bromo-5-iodotoluene ≥98%
2-Bromo-5-iodotoluene
Purity: 98+%
CAS No.: 202865-85-8
Molecular Formula: C7H6BrI
Formula Weight: 296.93
Refractive Index: 1.6490
Beilstein Reference: 8760525
Formula:
C₇H₆BrI MW: 296,93 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00070749 CAS Number: 202865-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...