2-Bromo-4-isopropylaniline ≥97%

Supplier: Thermo Fisher Scientific
Danger

A14995.14 A14995.06
ALFAA14995.14EA 0 SGD
ALFAA14995.14 ALFAA14995.06
2-Bromo-4-isopropylaniline ≥97%
2-Bromo-4-isopropylaniline

Purity: 97% CAS No.: 51605-97-1 Molecular Formula: C9H12BrN Formula Weight: 214.11 Boiling Point: 116-118°/3mm Beilstein Reference: 2638470 UN No.: 2810 MDL No.: MFCD00051585

Formula: C₉H₁₂BrN
MW: 214.1 g/mol
Boiling Pt: 116…118 °C (3 mmHg)
Density: 1.350
Storage Temperature: Ambient
MDL Number: MFCD00051585
CAS Number: 51605-97-1
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR