3-Bromo-o-toluidine ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Amino-6-bromotoluene, 3-Bromo-2-methylaniline
A11884.06
A11884.14
ALFAA11884.06EA
143
SGD
ALFAA11884.06
ALFAA11884.14
3-Bromo-o-toluidine ≥98%
3-Bromo-o-toluidine
Purity: 98+%
CAS No.: 55289-36-6
Molecular Formula: C7H8BrN
Formula Weight: 186.06
Flash Point:
Formula:
C₇H₈BrN MW: 186,05 g/mol Boiling Pt: 253…255 °C Density: 1,510 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051579 CAS Number: 55289-36-6 EINECS: 259-568-5 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...