3-Bromobenzyl alcohol ≥99%

Supplier: Thermo Fisher Scientific

Synonyms: (3-Bromophenyl)methanol

A10929.06 A10929.14
ALFAA10929.06EA 101 SGD
ALFAA10929.06 ALFAA10929.14
3-Bromobenzyl alcohol ≥99%
3-Bromobenzyl alcohol

Purity: 99% CAS No.: 15852-73-0 Molecular Formula: C7H7BrO Formula Weight: 187.04 Flash Point: 110°(230°F) Refractive Index: 1.5840 Boiling Point: 165°/16mm Beilstein Reference: 2242512 MDL No.: MFCD00004629

Formula: C₇H₇BrO
Boiling Pt: 165 °C (16 mmHg)
Density: 1,560
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004629
CAS Number: 15852-73-0
EINECS: 239-975-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR