2',4'-Dichloroacetophenone ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 2′,4′-Dichloroacetophenone

A14428.22 A14428.36
ALFAA14428.22EA 0 SGD
ALFAA14428.22 ALFAA14428.36
2',4'-Dichloroacetophenone ≥98%
2',4'-Dichloroacetophenone

Purity: 98% CAS No.: 2234-16-4 Molecular Formula: C8H6Cl2O Formula Weight: 189.04 Flash Point: 110°(230°F) Refractive Index: 1.5635 Melting Point: 31-34° Boiling Point: 245-247° Beilstein Reference: 637819 MDL No.: MFCD00000581

Formula: C₈H₆Cl₂O
MW: 189,04 g/mol
Boiling Pt: 245…247 °C
Melting Pt: 31…34 °C
Density: 1,320
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000581
CAS Number: 2234-16-4
EINECS: 218-780-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR