2'-Chloroacetophenone ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
1-(2-Chlorophenyl)ethanone, o-Chloroacetophenone
A15232.18
A15232.30
ALFAA15232.18EA
0
SGD
ALFAA15232.18
ALFAA15232.30
2'-Chloroacetophenone ≥98%
2'-Chloroacetophenone
Purity: 98%
CAS No.: 2142-68-9
Molecular Formula: C8H7ClO
Formula Weight: 154.60
Flash Point: 88°(190°F)
Refractive Index: 1.5435
Boiling Point: 228-229°
Beilstein Reference: 1858916
UN No.: 3416
MDL No.: MFCD00000560
Formula:
C₈H₇ClO MW: 154,6 g/mol Boiling Pt: 228…229 °C Density: 1,189 Flash Pt: 88 °C (190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000560 CAS Number: 2142-68-9 EINECS: 218-397-6 UN: 3416 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...