6-Chloro-m-toluic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
B25347.03
B25347.06
B25347.14
ALFAB25347.03EA
0
SGD
ALFAB25347.03
ALFAB25347.06
ALFAB25347.14
6-Chloro-m-toluic acid ≥98%
6-Chloro-m-toluic acid
Purity: 98%
CAS No.: 6342-60-5
Molecular Formula: C8H7ClO2
Formula Weight: 170.60
MDL No.: MFCD00045798
Formula:
C₈H₇ClO₂ MW: 170,6 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00045798 CAS Number: 6342-60-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...