3-Chloro-o-toluic acid ≥98%

Supplier: Thermo Scientific
Warning

L15773.03 L15773.06
ALFAL15773.03EA 0 SGD
ALFAL15773.03 ALFAL15773.06
3-Chloro-o-toluic acid ≥98%
3-Chloro-o-toluic acid

Purity: 98% CAS No.: 7499-08-3 Molecular Formula: C8H7ClO2 Formula Weight: 170.60 Melting Point: 162-166° Beilstein Reference: 1072826

Formula: C₈H₇ClO₂
MW: 170,6 g/mol
Melting Pt: 162…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00053312
CAS Number: 7499-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR