α-Chloro-2,6-difluorotoulene ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-(Chloromethyl)-1,3-difluorobenzene, 2,6-Difluorobenzyl chloride, 1-(Chloromethyl)-2,6-difluorobenzene

A19472.03 A19472.06 A19472.14
ALFAA19472.03EA 0 SGD
ALFAA19472.03 ALFAA19472.06 ALFAA19472.14
α-Chloro-2,6-difluorotoulene ≥98%
α-Chloro-2,6-difluorotoulene

Purity: 98% CAS No.: 697-73-4 Molecular Formula: C7H5ClF2 Formula Weight: 162.57 Melting Point: 32-35° Boiling Point: 172-173°/640mm UN No.: 1760 MDL No.: MFCD00191346

Formula: C₇H₅ClF₂
MW: 162.57 g/mol
Boiling Pt: 172…173 °C (640 mmHg)
Melting Pt: 32…35 °C
Storage Temperature: Ambient
MDL Number: MFCD00191346
CAS Number: 697-73-4
UN: 1760
ADR: 8 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR