α-Chloro-3,4,5-trifluorotoulene ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
1-(Chloromethyl)-3,4,5-trifluorobenzene, 3,4,5-Trifluorobenzyl chloride
L19647.03
L19647.06
L19647.14
ALFAL19647.03EA
0
SGD
ALFAL19647.03
ALFAL19647.06
ALFAL19647.14
α-Chloro-3,4,5-trifluorotoulene ≥98%
α-Chloro-3,4,5-trifluorotoulene
Purity: 98%
CAS No.: 732306-27-3
Molecular Formula: C7H4ClF3
Formula Weight: 180.56
Refractive Index: 1.4740
Formula:
C₇H₄ClF₃ MW: 180,56 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03001154 CAS Number: 732306-27-3 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...