2-Chloro-3,5-dinitro-α,α,α-trifluorotoluene ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 2-Chloro-1,5-dinitro-3-(trifluoromethyl)benzene, 2-Chloro-3,5-dinitrobenzotrifluoride, 1-Chloro-2,4-dinitro-6-(trifluoromethyl)benzene

A17617.06 A17617.14
ALFAA17617.06EA 0 SGD
ALFAA17617.06 ALFAA17617.14
2-Chloro-3,5-dinitro-α,α,α-trifluorotoluene ≥98%
2-Chloro-3,5-dinitro-α,α,α-trifluorotoluene

Purity: 98+% CAS No.: 392-95-0 Molecular Formula: C7H2ClF3N2O4 Formula Weight: 270.55 Melting Point: 61-64° Beilstein Reference: 2061156 UN No.: 2923 MDL No.: MFCD00007076

Formula: C₇H₂ClF₃N₂O₄
MW: 270,55 g/mol
Melting Pt: 61…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00007076
CAS Number: 392-95-0
EINECS: 206-883-0
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR