2-Chloro-6-iodotoluene ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 1-Chloro-3-iodo-2-methylbenzene

A11582.03 A11582.06 A11582.14
ALFAA11582.03EA 0 SGD
ALFAA11582.03 ALFAA11582.06 ALFAA11582.14
2-Chloro-6-iodotoluene ≥98%
2-Chloro-6-iodotoluene

Purity: 98% CAS No.: 42048-11-3 Molecular Formula: C7H6ClI Formula Weight: 252.48 Refractive Index: 1.6273 Melting Point: 11° Boiling Point: 119-120°/15mm Beilstein Reference: 2433704 MDL No.: MFCD00019013

Formula: C₇H₆ClI
MW: 252,48 g/mol
Boiling Pt: 119…120 °C (15 mmHg)
Melting Pt: 11 °C
Density: 1,820
Storage Temperature: Ambient
MDL Number: MFCD00019013
CAS Number: 42048-11-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR