3-Chloro-o-toluidine ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 2-Amino-6-chlorotoluene, 3-Chloro-2-methylaniline

A18221.0B A18221.30
ALFAA18221.0BEA 406 SGD
ALFAA18221.0B ALFAA18221.30
3-Chloro-o-toluidine ≥98%
3-Chloro-o-toluidine

Purity: 98+% CAS No.: 87-60-5 Molecular Formula: C7H8ClN Formula Weight: 141.60 Flash Point: 113°(235°F) Refractive Index: 1.5890 Melting Point: 2° Boiling Point: 245° Beilstein Reference: 471697 UN No.: 3429 MDL No.: MFCD00007766

Formula: C₇H₈ClN
MW: 141,6 g/mol
Boiling Pt: 245 °C
Melting Pt: 2 °C
Density: 1,188
Flash Pt: 113 °C (235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007766
CAS Number: 87-60-5
EINECS: 201-756-6
UN: 3429
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR