(3,4,5-Trifluorophenyl)acetonitrile ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-(3,4,5-Trifluorophenyl)acetonitrile, 3,4,5-Trifluorobenzyl cyanide
H26200.03
H26200.06
ALFAH26200.03EA
0
SGD
ALFAH26200.03
ALFAH26200.06
(3,4,5-Trifluorophenyl)acetonitrile ≥98%
(3,4,5-Trifluorophenyl)acetonitrile
Purity: 98%
CAS No.: 220228-03-5
Molecular Formula: C8H4F3N
Formula Weight: 171.12
Melting Point: 30-32°
UN No.: 3439
MDL No.: MFCD00083531
Formula:
C₈H₄F₃N MW: 171,12 g/mol Melting Pt: 30…32 °C Storage Temperature: Ambient |
MDL Number:
MFCD00083531 CAS Number: 220228-03-5 EINECS: None UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...