2',4'-Difluoropropiophenone ≥98%
Supplier: Thermo Scientific
Synonyms:
1-(2,4-Difluorophenyl)propan-1-one
A12032.06
A12032.14
ALFAA12032.06EA
0
SGD
ALFAA12032.06
ALFAA12032.14
2',4'-Difluoropropiophenone ≥98%
2',4'-Difluoropropiophenone
Purity: 98%
CAS No.: 85068-30-0
Molecular Formula: C9H8F2O
Formula Weight: 170.16
Flash Point: 76°(168°F)
Refractive Index: 1.4850
Boiling Point: 51-52°/1.5mm
Beilstein Reference: 7633370
MDL No.: MFCD00015507
Formula:
C₉H₈F₂O MW: 170,16 g/mol Boiling Pt: 51…52 °C (1,5 mmHg) Density: 1,193 Flash Pt: 76 °C (168 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015507 CAS Number: 85068-30-0 EINECS: 285-291-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...