3',4',5'-Trifluoroacetophenone ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 1-(3,4,5-Trifluorophenyl)ethan-1-one

L19187.03 L19187.06
ALFAL19187.03EA 0 SGD
ALFAL19187.03 ALFAL19187.06
3',4',5'-Trifluoroacetophenone ≥97%
3',4',5'-Trifluoroacetophenone

Purity: 97% CAS No.: 220141-73-1 Molecular Formula: C8H5F3O Formula Weight: 174.12 Flash Point: 100°(212°F) Refractive Index: 1.4730 MDL No.: MFCD00083533

Formula: C₈H₅F₃O
MW: 174,12 g/mol
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00083533
CAS Number: 220141-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR