3'-Fluoropropiophenone ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
m-Fluoropropiophenone, 1-(3-Fluorophenyl)propan-1-one
L11922.03
L11922.06
ALFAL11922.03EA
0
SGD
ALFAL11922.03
ALFAL11922.06
3'-Fluoropropiophenone ≥97%
3'-Fluoropropiophenone
Purity: 97%
CAS No.: 455-67-4
Molecular Formula: C9H9FO
Formula Weight: 152.17
Refractive Index: 1.5050
Boiling Point: 94-96°/5mm
MDL No.: MFCD00061245
Formula:
C₉H₉FO MW: 152,17 g/mol Boiling Pt: 94…96 °C (5 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00061245 CAS Number: 455-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...