3-Iodo-p-toluic acid ≥98%

Supplier: Thermo Fisher Scientific
Warning

A14197.06 A14197.14
ALFAA14197.06EA 0 SGD
ALFAA14197.06 ALFAA14197.14
3-Iodo-p-toluic acid ≥98%
3-Iodo-p-toluic acid

Purity: 98% CAS No.: 82998-57-0 Molecular Formula: C8H7IO2 Formula Weight: 262.05 Melting Point: 206-210° Beilstein Reference: 2436216

Formula: C₈H₇IO₂
MW: 262.05 g/mol
Boiling Pt: 329
Melting Pt: 206…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00010392
CAS Number: 82998-57-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR