2,6-Dimethoxytoluene ≥98%

Supplier: Thermo Scientific
Warning

B22606.14 B22606.22
ALFAB22606.14EA 0 SGD
ALFAB22606.14 ALFAB22606.22
2,6-Dimethoxytoluene ≥98%
2,6-Dimethoxytoluene

Purity: 98% CAS No.: 5673-07-4 Molecular Formula: C9H12O2 Formula Weight: 152.19 Flash Point: 96°(204°F) Odour: Aromatic Boiling Point: 222° Beilstein Reference: 2045327 MDL No.: MFCD00008374

Formula: C₉H₁₂O₂
MW: 152,19 g/mol
Boiling Pt: 222 °C
Melting Pt: 37…41 °C
Flash Pt: 96 °C (204 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008374
CAS Number: 5673-07-4
EINECS: 227-131-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR