3,4,5-Trimethoxytoluene ≥98%
Supplier: Thermo Scientific
Warning
|
Synonyms:
1,2,3-Trimethoxy-5-methylbenzene
A15201.06
A15201.14
A15201.18
ALFAA15201.06EA
0
SGD
ALFAA15201.06
ALFAA15201.14
ALFAA15201.18
3,4,5-Trimethoxytoluene ≥98%
3,4,5-Trimethoxytoluene
Purity: 98%
CAS No.: 6443-69-2
Molecular Formula: C10H14O3
Formula Weight: 182.22
Flash Point: 74°(165°F)
Refractive Index: 1.5230
Melting Point: 34-38°
Boiling Point: 115-116°/4mm
Beilstein Reference: 1953791
MDL No.: MFCD00008397
Formula:
C₁₀H₁₄O₃ MW: 182,22 g/mol Boiling Pt: 115…116 °C (4 mmHg) Melting Pt: 34…38 °C Density: 1,082 Flash Pt: 74 °C (165 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008397 CAS Number: 6443-69-2 EINECS: 229-239-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...