2-Ethylthiophenol 95%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Ethylbenzene-1-thiol, 1-Ethyl-2-mercaptobenzene, 2-Ethylbenzenethiol
L11498.09
L11498.04
ALFAL11498.09EA
212
SGD
ALFAL11498.09
ALFAL11498.04
2-Ethylthiophenol 95%
2-Ethylthiophenol
Purity: 95%
CAS No.: 4500-58-7
Molecular Formula: C8H10S
Formula Weight: 138.23
Flash Point: 80°(176°F)
Refractive Index: 1.5690
Odour: Stench
Boiling Point: 203-205°
Beilstein Reference: 2553288
UN No.: 2810
MDL No.: MFCD00010022
Formula:
C₈H₁₀S MW: 138,23 g/mol Boiling Pt: 203…205 °C Density: 1,025 Flash Pt: 80 °C (176 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00010022 CAS Number: 4500-58-7 EINECS: 224-811-6 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...