3-Benzyloxybenzyl alcohol 97

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (3-(Benzyloxy)phenyl)methanol

H27707.14 H27707.06
ALFAH27707.14EA 0 SGD
ALFAH27707.14 ALFAH27707.06
3-Benzyloxybenzyl alcohol 97
3-Benzyloxybenzyl alcohol

Purity: 97% CAS No.: 1700-30-7 Molecular Formula: C14H14O2 Formula Weight: 214.26 Flash Point: 110°(230°F) Melting Point: 47-51° MDL No.: MFCD00004642

Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
Boiling Pt: 160 °C (3 mmHg)
Melting Pt: 47…51 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004642
CAS Number: 1700-30-7
EINECS: 216-931-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR