3,4-Dimethylbenzamide ≥98%
Supplier: Thermo Scientific
|
Warning
|
A11995.03
A11995.06
A11995.14
ALFAA11995.03EA
48.7
SGD
ALFAA11995.03
ALFAA11995.06
ALFAA11995.14
3,4-Dimethylbenzamide ≥98%
3,4-Dimethylbenzamide
Purity: 98%
CAS No.: 5580-33-6
Molecular Formula: C9H11NO
Formula Weight: 149.19
Melting Point: 101-103°
MDL No.: MFCD00052960
Formula:
C₉H₁₁NO Melting Pt: 101…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052960 CAS Number: 5580-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...