2,6-Diisopropylphenyl isothiocyanate 97%

Supplier: Thermo Scientific
Danger

Synonyms: 1,3-Diisopropyl-2-isothiocyanatobenzene

L10893.03 L10893.06
ALFAL10893.03EA 81.9 SGD
ALFAL10893.03 ALFAL10893.06
2,6-Diisopropylphenyl isothiocyanate 97%
2,6-Diisopropylphenyl isothiocyanate

Purity: 97% CAS No.: 25343-70-8 Molecular Formula: C13H17NS Formula Weight: 219.35 Flash Point: 110°(230°F) Refractive Index: 1.5830 Boiling Point: 139-140°/5mm Beilstein Reference: 2416576 UN No.: 2922 MDL No.: MFCD00041341

Formula: C₁₃H₁₇NS
MW: 219,35 g/mol
Boiling Pt: 139…140 °C (5 mmHg)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041341
CAS Number: 25343-70-8
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR