2,6-Xylyl isothiocyanate ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 2,6-Dimethylphenylisothiocyanate, 2,6-Dimethylphenyl isothiocyanate

A19480.06 A19480.14
ALFAA19480.06EA 0 SGD
ALFAA19480.06 ALFAA19480.14
2,6-Xylyl isothiocyanate ≥98%
2,6-Xylyl isothiocyanate

Purity: 98% CAS No.: 19241-16-8 Molecular Formula: C9H9NS Formula Weight: 163.24 Flash Point: 110°(230°F) Refractive Index: 1.6250 Odour: Pungent Boiling Point: 247° Beilstein Reference: 637195 UN No.: 2922 MDL No.: MFCD00038715

Formula: C₉H₉NS
MW: 163,24 g/mol
Boiling Pt: 247 °C
Density: 1,085
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00038715
CAS Number: 19241-16-8
EINECS: 242-906-0
UN: 2922
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR