3,5-Xylyl isothiocyanate ≥99%

Supplier: Thermo Scientific
Danger

Synonyms: 3,5-Dimethylphenyl isothiocyanate, 3,5-Dimethylphenylisothiocyanate

A11302.06 A11302.14
ALFAA11302.06EA 145 SGD
ALFAA11302.06 ALFAA11302.14
3,5-Xylyl isothiocyanate ≥99%
3,5-Xylyl isothiocyanate

Purity: 99% CAS No.: 40046-30-8 Molecular Formula: C9H9NS Formula Weight: 163.24 Refractive Index: 1.6190 Beilstein Reference: 6856812 UN No.: 2922 MDL No.: MFCD00041082

Formula: C₉H₉NS
MW: 163,24 g/mol
Boiling Pt: 261 °C (1013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00041082
CAS Number: 40046-30-8
UN: 2922
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR