2-Hydroxyethylmesitylene ≥98%

Supplier: Thermo Scientific

Synonyms: 1-(2-Hydroxyethyl)-2,4,6-trimethylbenzene, 2-(2,4,6-Trimethylphenyl)ethanol, 2-Hydroxyethylmesitylene, 2,4,6-Trimethylphenethyl alcohol, 2-Mesitylethanol

L08032.06 L08032.03
ALFAL08032.06EA 0 SGD
ALFAL08032.06 ALFAL08032.03
2-Hydroxyethylmesitylene ≥98%
2-Hydroxyethylmesitylene

Purity: 98% CAS No.: 6950-92-1 Molecular Formula: C11H16O Formula Weight: 164.25 Melting Point: 80-84° Beilstein Reference: 1938400

Formula: C₁₁H₁₆O
MW: 164,25 g/mol
Melting Pt: 80…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00020609
CAS Number: 6950-92-1
EINECS: 230-123-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR